Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
Brand:  Epredia™ 1220
 View more versions of this product
Product Code. 11919671
Description
Description
  • Constructed of rigid transparent plastic.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
22x22 to 12x12 mm, 20 mm deep
Truncated, tapered
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Peel-A-Way™ Disposable Embedding Molds > 0.47 x 0.47 x 0.79 in. (12 x 12 x 20mm)