missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Multi-Spot Slides
Wash and reuse Epredia™ Shandon™ Multi-Spot Slides, featuring a coating that allows for several cleanings.
Brand: Epredia™ 9991090
This item is not returnable.
View return policy
Specifications
76 mm | |
Glass | |
Plain | |
6 mm Wells |
26 mm | |
Multispot Slide | |
6mm Wells | |
6 mm |