missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin

Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.

Brand:  Epredia™ 9990918

 View more versions of this product

Product Code. 12590057

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available
Specifications

Specifications

125/Cs.
Formalin
120mL specimen container filled to 60mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers