missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Shandon™ EZ-Mount™ Mountant
Inhibit staint fading with Thermo Scientific™ Shandon™ EZ-Mount™ Mountant, a clear, xylene-based mounting medium compatible with membrane filters.
Brand: Thermo Scientific™ 9999120
This item is not returnable.
View return policy
Description
Inhibit staint fading with Thermo Scientific™ Shandon™ EZ-Mount™ Mountant, a clear, xylene-based mounting medium compatible with membrane filters.
- Refractive index: 1.495 ± 0.005 (dry)
- Viscosity: High (1280-2790 cps at 22°C)
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Reproductive toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
- H360-May damage fertility or the unborn child.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P271-Use only outdoors or in a well-ventilated area.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P363-Wash contaminated clothing before reuse.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : xylenes, butyl benzyl phthalate, 2-propenoic acid, 2-methyl-butyl ester, polymereith methyl-2-methy