Learn More
Epredia™ Shandon™ EA-65 Stain
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.
76.90 € - 144.00 €
Specifications
Type Cellular Components Stain
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12519996
View Documents
Epredia™
6766003
2 x 1L
76.90 €
Pack of 2
12647936
View Documents
Epredia™
6766004
4L
144.00 €
Each
Description
Description
  • Polychromatic cytoplasmic stain
  • Ideal for thicker non-GYN preparations
  • Biological Stain Commission-certified dye
Specifications
Specifications
Cellular Components Stain