Learn More
SGTB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15895-1-AP-20UL
 View more versions of this product
Product Code. 17541336
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The SGTB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SGTB. This antibody recognizes human, mouse, rat antigen. The SGTB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SGTB
Polyclonal
Unconjugated
20 μL
54557
IgG
Western Blot
133 μg/mL
Rabbit
Primary
Human, Mouse, Rat