missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGSH Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | SGSH |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886015
|
Proteintech
28150-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876015
|
Proteintech
28150-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The SGSH antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SGSH. This antibody recognizes human antigen. The SGSH antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
SGSH | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
6448 | |
Primary |