missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24654-1-AP-150UL
This item is not returnable.
View return policy
Description
Sodium-dependent glucose transporter. Has a Na+ to glucose coupling ratio of 1:1. Efficient substrate transport in mammalian kidney is provided by the concerted action of a low affinity high capacity and a high affinity low capacity Na+/glucose cotransporter arranged in series along kidney proximal tubules.Specifications
SGLT2 | |
Polyclonal | |
Unconjugated | |
SLC5A2 | |
Na(+)/glucose cotransporter 2, SGLT2, SLC5A2, Sodium/glucose cotransporter 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246787, 6524 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31639, Q923I7 | |
SLC5A2 | |
SGLT2 Fusion Protein Ag19163 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |