Learn More
Trajan™ SilTite™ FingerTite Blanking Ferrule
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073633
Product Code. 15248983
  • 34.70 € / Pack of 2
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Blank Ferrule
SilTite
2/Pk.