missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ eVol™ XCHANGE Syringes
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
96.50 € - 259.00 €
Specifications
For Use With (Application) | Liquid handling procedure |
---|---|
Length (English) | 1.9 in. |
Length (Metric) | 50 mm |
Product Type | eVol™ Syringe |
Length (Metric) Needle | 50 mm |
Product Code | Brand | Volume (Metric) | Needle Point Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Point Style | Price | Quantity & Availability | ||||
12977586
|
Trajan™
2910029 |
100 μL | Bevel |
96.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13467878
|
Trajan™
2910035 |
1 mL | Bevel |
96.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12947596
|
Trajan™
2910329 |
100 μL | Bevel |
259.00 €
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Liquid handling user independent device
- Reduction in need for re-analysis
- Syringes easily and quickly changed
- Replacement parts available
Specifications
Liquid handling procedure | |
50 mm | |
50 mm |
1.9 in. | |
eVol™ Syringe | |
Bevel |