Learn More
SGE™ eVol™ NMR Starter Kit
User-calibrated analytical syringe improves accuracy and reproducibility over standard laboratory tools
Brand: SGE™ 2910100
This item is not returnable.
View return policy
Description
- The programmable digital drive means liquid handling procedures are user-independent, allowing more efficient workflow scheduling and a reduction in the re-analysis of incorrectly processed or false positive samples
- The gravimetric calibration procedure is simple, intuitive and can be performed at appropriate intervals
- Kit includes unit, charger, stand, 3 syringes: 5μL with 11.5cm needle, 50μL with 11.5cm and 18cm needles, and 500μL with 11.5cm and 18cm needles
Specifications
For user-calibrated analytical syringe improves accuracy and reproducibility over standard laboratory tools | |
Stand, universal charger, comprehensive, instruction manual, disc with manual in multiple languages, 3 syringes, 5μL syringe with 115mm needle, 50μL syringe with 115mm and 180mm needles, 500μL syringe | |
Electronic Syringe Starter Kit |