Learn More
Trajan™ Diamond Thermo RSH 100μL GC Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™ 0355611
 View more versions of this product
Product Code. 13127946
  • 106.00 € / Each
Explore more special offers
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
23 G
0.24 mm
0.63 mm
2.24 in.
1 Pk.
Ultra Smooth Hydrophilic, Cone
0.009 in.
0.024 in.
Gas chromatography
Autosampler Syringe
100 μL
CTC PAL GC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan™ Diamond Thermo RSH 100μL GC Autosampler Syringes > With Ultra Smooth Hydrophilic needle