Learn More
Trajan™ Diamond LC Needles for 100 to 500μL Manual and Autosampler Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.
Brand: Trajan™ 0355211
This item is not returnable.
View return policy
Description
Improve the sensitivity of your MS analysis:
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Stainless Steel | |
0.0066 in. | |
0.17 mm | |
Ultra Smooth Hydrophilic Coating | |
Diamond Needle | |
Manual and Autosampler Syringes (100 to 500μL) |
22 G | |
0.02 in. | |
0.72 mm | |
2 in. | |
LC |