Learn More
Trajan™ Diamond CTC PAL 10 to 50μL GC Autosampler Syringes
Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™
 View more versions of this product
Product Code. 13177936
  • 115.00 € / Each
Explore more special offers
Description
Description
  • Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
  • Ultra Smooth needles reduce non specific interaction between your sample and the needle. 
  • Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications
Specifications
23 G
0.11 mm
0.63 mm
Ultra Smooth Hydrophobic Coating
GC Autosampler Syringe
10 to 50 μL
CTC PAL
0.004 in.
0.024 in.
Gas chromatography
1.96 in.
Ea.
Cone