Learn More
Trajan™ Beckman/Altex, Rheodyne, SSI, Valco Valve Replacement Needle
Removable needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves
Brand:  Trajan™ 002313
Product Code. 15218453
  • 77.10 € / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Specifications
PTFE (Plunger)
22 G
0.028 mm
LC
Gas tight
0.17 mm
10 μL