missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS17A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | SFRS17A |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801664
|
Proteintech
13441-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16891654
|
Proteintech
13441-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.Specifications
SFRS17A | |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q02040 | |
8227 | |
SFRS17A Fusion Protein Ag4252 | |
Primary | |
-20°C | |
AKAP17A |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
721P, B lymphocyte antigen, CCDC133, CXYorf3, DXYS155E, Protein XE7, SFRS17A, XE7, XE7Y | |
AKAP17A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |