Learn More
Cytiva Serum Capsule with Versapor™ Membrane
Capsule for prefiltration of particule-laden samples, serum filtration, and dissolution testing.
Brand:  Cytiva 12168
Product Code. 17114231
  • 164.00 € / Each
Explore more special offers
Description
Description
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrylic Copolymer (Hydrophilic)
1.2 μm
Sterile