Learn More
SERPINA6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12010-1-AP-150UL
 View more versions of this product
Product Code. 16829463
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Specifications
Specifications
SERPINA6
Polyclonal
Unconjugated
SERPINA6
CBG, Serpin A6, SERPINA6, Transcortin
Rabbit
Antigen Affinity Chromatography
RUO
866
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08185
SERPINA6
SERPINA6 Fusion Protein Ag2623
150 μL