Learn More
Ohaus™ Series MB Sample Cage
OHAUS™ Series MB Sample Cage is suitable for use with MB90 AM, MB120 AM, MB27 AM, and MB23 AM Moisture Analyzers.
Brand: Ohaus™ 80252477
37.64 EUR valid until 2024-09-27
Use promo code "21704" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- For use with moisture analyzer
Specifications
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
0.540 kg |