missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Vinyl Powdered Blue Disposable Gloves

Product Code. 12565876
Click to view available options
Size: Medium
This item is not returnable. View return policy
This item is not returnable. View return policy

Disposable protective glove made of Vinyl is latex free. Semperguard™ Vinyl Powdered Blue Disposable Gloves contain no latex protiens or accelerators.

  • Very skin-friendly glove for people with latex allergies: contains no latex proteins or accelerators
  • Tested quality and use of selected raw materials
  • High quality vinyl formula provides excellent wearer comfort
  • Produced according to medical directives EN 455
  • Signal color blue is suitable for areas subject to HACCP standards
  • Tested and certified according to 1935/2004 - suitable for nonfat food contact

Specifications

Certifications/Compliance CE Category I, EN 420, EN 374, EN 455
Classification CE category I Protective glove for minimal risks
Color Blue
Disposable Single-use
Exterior Finish Smooth
Grade Suitable for areas subject to HACCP standards
Hand Ambidextrous
Latex Free Yes
Material Vinyl
Powdered Powdered
Product Type Disposable Gloves
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.14 mm
For Use With (Application) Suitable for nonfat food contact
Glove Cuff Style Rolled
Packaging Type Dispenser Box
Quantity 100/Bx
Tensile Strength 11 MPa
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.