missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Sempercare Shine+ Nitrile Examination Gloves

Product Code. 15809057
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A longer cuff offers users additional safety by covering wrists and underarms. Includes innovative foil opening for considerably easier glove removal.

  • The optimized length of the fingers significantly improves the wearing comfort
  • Powder-free natural latex examination glove excels by its extraordinary, well-gliding surface.
  • The textured fingers provide a good grip which is indispensable for safe instrument handling.
  • The synthetic inner coating achieved by the special Sempermed technology makes it easy and fast to don.

Specifications

Certifications/Compliance ASTM, EN 455, EN 420, EN 374-1, EN 374-3
Classification CE Class I, CE Category III
Color White
Disposable Single-use
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
Thickness (Metric) 0.15 mm
For Use With (Application) Hospital
Glove Cuff Style Extended
Inspected for Medical Use Yes
Packaging Type Dispenser Box
Quantity 200/Pk.
Resistant To Chemical
Length (Metric) 26.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.