missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Protects Hygienic Blue Nitrile Disposable Gloves

Product Code. 15363804
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder-free to reduce the risk of spreading germs and allergens. Semperit™ Semperguard™ Protects Hygienic Blue Nitrile Disposable Gloves have been developed especially for food and cleaning applications.

  • Sapphire blue nitrile to avoid latex allergies
  • Ambidextrous gloves with rim and textured fingers
  • Processed in line with European standard as per reg. 1935/2004

Specifications

Certifications/Compliance ISO 9001, EN 420, EN 374-2
Color Blue
Disposable Single-use
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (Metric) 0.11 mm
Finger Style Textured
For Use With (Application) Food and cleaning
Glove Cuff Style Rolled
Packaging Type Dispenser Box
Quantity 200/Pk.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.