missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Protects Hygienic Blue Nitrile Disposable Gloves

Product Code. 15353804
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder-free to reduce the risk of spreading germs and allergens. Semperit™ Semperguard™ Protects Hygienic Blue Nitrile Disposable Gloves have been developed especially for food and cleaning applications.

  • Sapphire blue nitrile to avoid latex allergies
  • Ambidextrous gloves with rim and textured fingers
  • Processed in line with European standard as per reg. 1935/2004

Specifications

Certifications/Compliance ISO 9001, EN 420, EN 374-2
Color Blue
Disposable Single-use
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.11 mm
Finger Style Textured
For Use With (Application) Food and cleaning
Glove Cuff Style Rolled
Packaging Type Dispenser Box
Quantity 200/Pk.
Length (Metric) 23 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'