Learn More
Selenium powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
231.15 € - 791.94 €
Chemical Identifiers
CAS | 7782-49-2 |
---|---|
Molecular Formula | Se |
Molecular Weight (g/mol) | 78.97 |
MDL Number | MFCD00134090 MFCD00011224 |
InChI Key | BUGBHKTXTAQXES-UHFFFAOYSA-N |
Synonym | selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray |
PubChem CID | 6326970 |
ChEBI | CHEBI:27568 |
IUPAC Name | selenium |
SMILES | [Se] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11373248
|
Thermo Scientific Alfa Aesar
036208.18 |
50 g |
231.15 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363248
|
Thermo Scientific Alfa Aesar
036208.30 |
250 g |
791.94 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-49-2 | |
78.97 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
6326970 | |
selenium |
Se | |
MFCD00134090 MFCD00011224 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
CHEBI:27568 | |
[Se] |
Specifications
7782-49-2 | |
99.999% (metals basis) | |
MFCD00134090 MFCD00011224 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
[Se] | |
78.97 | |
CHEBI:27568 | |
(metals basis) |
Powder | |
Se | |
UN3077 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
selenium | |
6326970 | |
99.999% | |
Selenium |
Safety and Handling
GHS H Statement
H301-H331-H373
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 231-957-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.