Learn More
Selenic acid, 40% aq. soln., Thermo Scientific Chemicals
273.36 € - 1044.20 €
Chemical Identifiers
CAS | 7783-08-6 |
---|---|
MDL Number | MFCD00012191 |
Synonym | selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2 |
ChEBI | CHEBI:18170 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353497
|
Thermo Scientific Alfa Aesar
018851.22 |
100 g |
273.36 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343497
|
Thermo Scientific Alfa Aesar
018851.36 |
500 g |
1044.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-08-6 | |
MFCD00012191 | |
selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2 | |
CHEBI:18170 |
Specifications
7783-08-6 | |
Liquid | |
H2SeO4 | |
UN2922 | |
selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2 | |
YAZJAPBTUDGMKO-UHFFFAOYSA-L | |
selenic acid | |
1089 | |
144.97 (Anhydrous) | |
1.41 g/mL |
Colorless | |
K2O4Se | |
MFCD00012191 | |
14,8429 | |
Miscible with sulfuric acid. Immiscible with ammonia. | |
O[Se](=O)(=O)O | |
221.16 | |
CHEBI:18170 | |
40% aq. soln. | |
Selenic acid |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 231-979-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only