missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Sedimentation Tube
Sedimentation Tube constructed of polystyrene features a conical shaped bottom.
Brand: BRAND™ 114740
166.97 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polystyrene constuction
- Clear
Specifications
105 mm | |
Polystyrene | |
16 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction