missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Sealing Sleeve Set
Sampling Systems™ Sealing Sleeve Set is designed to enable a pocket sampler to be emptied.
15.90 € - 27.40 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15859391
|
Sampling Systems™
8005A-01 |
25 mm |
15.90 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15869391
|
Sampling Systems™
8005A-02 |
40 mm |
27.40 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
- Enables one pocket to be emptied at a time
- Simply clip on to cover
- Easy to remove
Specifications
Polyethylene | |
Set of 3 sleeves for sampler |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title