missing translation for 'onlineSavingsMsg'
Learn More
Ansell™ Scorpio™ 19-024 Series Black Neoprene Immersion Gloves Product Code.: 11751734
In Stock
to see stock.
Add to basket

Ansell™ Scorpio™ 19-024 Series Black Neoprene Immersion Gloves

Product Code. 11751734 Shop All Ansell Products
10
Click to view available options
Size:
8
10
This item is not returnable. View return policy

Product Code. 11751734

Brand: Ansell™ 19024100

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Combine the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner. Provides protection from intermittent contact with hot surfaces up to 180°C.

  • Chemical resistant gloves
  • Combines the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner
  • Designed to protect the users in applications where thermal and chemical risks are present
  • Good mechanical and chemical properties
  • Unaffected by detergents and cleaning solutions
  • Offers protection against greases and oils
  • Superior flexibility and grip

Applications:

  • Charging and blending raw materials
  • Handling materials with sharp or rough edges
  • Press work
  • Production line support and maintenance
  • Unexpected leakages spills or other releases
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 511:2006 , EN388:2003 , EN 407:2004, EN 374:2003
Color Black
Exterior Finish Rough
Latex Free Yes
Material Neoprene
Product Type Chemical and Liquid Protection Gloves
Size 10
Glove Cuff Style Gauntlet
Liner Material Double insulating liner
Packaging Type 1 pair in a bag; 12 bags in a carton
Series 19-024
Length (Metric) 450 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Scorpio™ 19-024 Series Black Neoprene Immersion Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.