Learn More
SCNM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15490-1-AP-20UL
 View more versions of this product
Product Code. 16834694
  • 145.00 € / 20µL
Explore more special offers
Description
Description
SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.
Specifications
Specifications
SCNM1
Polyclonal
Unconjugated
SCNM1
SCNM1, sodium channel modifier 1
Rabbit
Antigen Affinity Chromatography
RUO
310662, 69269, 79005
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K136, Q9BWG6
SCNM1
SCNM1 Fusion Protein Ag7854
20 μL
Primary
Human, Rat, Mouse