missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23393-1-AP-20UL
This item is not returnable.
View return policy
Description
Stearoyl-CoA desaturase., ,Specifications
SCD | |
Polyclonal | |
Unconjugated | |
SCD | |
Acyl CoA desaturase, Delta(9) desaturase, FADS5, Fatty acid desaturase, MSTP008, SCD, SCD1, SCDOS, Stearoyl CoA desaturase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246074, 6319 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00767 | |
SCD | |
SCD Fusion Protein Ag13456 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction