Learn More
Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals
310.00 € - 1230.00 €
Chemical Identifiers
CAS | 14474-33-0 |
---|---|
Molecular Formula | I3Sc |
Molecular Weight (g/mol) | 425.669 |
MDL Number | MFCD00054037 |
InChI Key | HUIHCQPFSRNMNM-UHFFFAOYSA-K |
Synonym | scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e |
PubChem CID | 84464 |
IUPAC Name | scandium(3+);triiodide |
SMILES | [Sc+3].[I-].[I-].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321299
|
Thermo Scientific Alfa Aesar
013687.03 |
1 g |
310.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331299
|
Thermo Scientific Alfa Aesar
013687.06 |
5 g |
1230.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Scandium iodide is used in high-intensity lighting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
14474-33-0 | |
425.669 | |
HUIHCQPFSRNMNM-UHFFFAOYSA-K | |
84464 | |
[Sc+3].[I-].[I-].[I-] |
I3Sc | |
MFCD00054037 | |
scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e | |
scandium(3+);triiodide |
Specifications
945°C | |
Powder | |
MFCD00054037 | |
Soluble in water. | |
[Sc+3].[I-].[I-].[I-] | |
425.669 | |
425.67 | |
Ampule | |
(REO) |
14474-33-0 | |
I3Sc | |
scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e | |
HUIHCQPFSRNMNM-UHFFFAOYSA-K | |
scandium(3+);triiodide | |
84464 | |
99.999% | |
Hygroscopic | |
Scandium(III) iodide, Ultra dry |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-469-0
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only