missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 202190, 99+%, Thermo Scientific Chemicals
Inhibits p38 kinase activity in vivo through competition with ATP
146.00 € - 416.00 €
Chemical Identifiers
CAS | 152121-30-7 |
---|---|
Molecular Formula | C20H14FN3O, C20H14FN3O |
Synonym | 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463629
|
Thermo Scientific Alfa Aesar
J60950.MB |
25 mg |
146.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483629
|
Thermo Scientific Alfa Aesar
J60950.MF |
50 mg |
269.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473629
|
Thermo Scientific Alfa Aesar
J60950.MC |
100 mg | |||||||
Description
Inhibits p38 kinase activity through competition with ATP
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
152121-30-7 | |
C20H14FN3O, C20H14FN3O | |
4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole |
Specifications
Yellow | |
Inhibits p38 kinase activity in vivo through competition with ATP | |
C20H14FN3O, C20H14FN3O | |
4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole | |
Soluble in DMSO at 30mg/ml | |
Crystalline solid | |
FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1 | |
331.35 |
-30°C to -10°C | |
152121-30-7 | |
331.35g/mol | |
240°C to 243°C | |
Signal Transduction Reagent | |
NJNKPVPFGLGHPA-UHFFFAOYSA-N | |
4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one | |
≥99% |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only