missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sterile Petri Dishes
Choose 60mm (14-555-734) or 90mm (14-555-735) diameter dishes sterile disposable petri dishes
65.25 € - 75.45 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10456551
|
Sartorius
14311--60------N |
60 mm |
65.25 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10720403
|
Sartorius
14311--90------N |
90 mm |
75.45 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- Disposable
- Type 143
- Packaging: auto-sterile
Specifications
Single-use | |
Sterile | |
Auto-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title