Learn More
Sartorius Microsart™ Mini Vacuum Pumps
Microsart Mini vacuum pumps provide up-to-date technology for daily use in the microbiology laboratory environment
Brand:  Sartorius 16694-1-60-06
Product Code. 15952540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ideal for microbiological filtrations
  • Neoprene membrane pumps
  • Decreased sub lethals, higher recovery rates and shorter incubation times
  • Low noise level
  • Oil-and maintenance-free
  • Controlled and easily adjustable vacuum
  • Pressure filtration with Microsart mini vac possible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4.18 lb.
0.63 A
6.5 x 3.5 x 5.6 in. (164 x 90 x 141 mm)
65 w
40°C
60 Hz
14.2 cm
16.4 cm
90 mm
115 V
1.9 kg
115 V 60 Hz
53.5 dB (A)
104°F
6 L/min.
5.6 in.
6.5 in.
3.5 in.
2.5 bar