missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab P20
BIOPHARMA

Pressure filtration unit for sterile filtration or clarification of media, aqueous solutions and solvent mixtures in batches ranging from 100 ml to 5 L.

Brand:  Sartorius 18090----------D

Product Code. 18518074

  • 116.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • It features a female luer-lock inlet and a male luer-lock outlet
  • Highest flow rates with a large surface of filtration (20 cm²)
  • No loss of protein with a low binding membrane
  • Low dead volume due to an optimized membrane support
  • Versions available with a prefilter for high particle load solutions
  • Sartolab P20 Plus with an incorporated prefilter is recommended for difficult-to-filter solutions such as media that contains serum
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Sartolab P20
PES
5 L
20 cm²
1 mL
Ready-to-Use Pressure Filter
Polycarbonate
0.2 μm
Sterile
Pressure filter
61 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers