missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saponin, From Saponegin Glycosides (Off-White Powder/Practical Grade), MP Biomedicals
Plant origin, Saponegin glycosides
83.90 € - 769.00 €
Chemical Identifiers
CAS | 637-39-8 |
---|---|
Molecular Formula | C6H16ClNO3 |
Molecular Weight (g/mol) | 185.648 |
InChI Key | HHLJUSLZGFYWKW-UHFFFAOYSA-N |
Synonym | triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy |
PubChem CID | 101814 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
SMILES | C(CO)N(CCO)CCO.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11496222
|
MP Biomedicals
0210285525 |
25 g |
83.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406232
|
MP Biomedicals
0210285580 |
100 g |
187.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416232
|
MP Biomedicals
0210285590 |
500 g |
769.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
637-39-8 | |
185.648 | |
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
C6H16ClNO3 | |
HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
101814 | |
C(CO)N(CCO)CCO.Cl |
Specifications
637-39-8 | |
C6H16ClNO3 | |
HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride | |
101814 | |
Powder |
White | |
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
C(CO)N(CCO)CCO.Cl | |
185.648 | |
Practical | |
Saponin, Cell biology |
Safety and Handling
missing translation for 'einecsNumber' : 232-462-6
Recommended Storage : Store at room temperature (15° to 30°C)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Saponin, Practical Grade
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title