Learn More
Sand, washed, Thermo Scientific Chemicals
40.80 € - 138.00 €
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15477837
|
Thermo Scientific Alfa Aesar
H37933.0B |
1000 g |
40.80 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487837
|
Thermo Scientific Alfa Aesar
H37933.0I |
5000 g |
138.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230°C | |
14808-60-7 | |
Granules | |
SiO2 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 | |
Sand, Washed |
1710°C | |
Brown-Cream | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
Insoluble in water. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Odorless |
Safety and Handling
GHS H Statement
H350-H373
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P281-P308+P313-P501c
H350
missing translation for 'einecsNumber' : 238-878-4
missing translation for 'rtecsNumber' : W7330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only