missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ SampleSafe™ Seal
Buerkle™ SampleSafe™ Seal feature a one-way tightening mechanism that offers additional security against manipulation.
Brand: Buerkle™ 5399-2902
95.09 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Clear identification via numbers in sequence
- 100 seals (10 strips) per pack
- Protection against manipulation by stainless steel safety spring
Specifications
Red | |
Numers in sequence, stainless steel safety spring | |
260 mm | |
1.76 lb. | |
Seal |
Retention Samples | |
10.23 in. | |
100/Pk. | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction