missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Sample Ejection Rod

For use with Powder Lance cross-sectional samplers for cohesive powders.

Brand:  Sampling Systems™ 8570A-1100

 View more versions of this product

Product Code. 15859521

  • 155.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from 316L stainless steel
  • Push the sample ejection rod into the Powder Lance to push out the sample
  • Aids in sampling large volumes of cohesive powders
  • Available in two different lengths
Specifications

Specifications

Sample Ejection Rod
Sampling large volumes of cohesive powders
1100 mm
Stainless Steel (316L)
Powder Lance cross-sectional sampler
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers