Learn More
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
81.10 € - 213.00 €
Chemical Identifiers
CAS | 13759-83-6 |
---|---|
Molecular Formula | H12N3O15Sm |
Molecular Weight (g/mol) | 444.46 |
MDL Number | MFCD00149857 |
InChI Key | HDCOFJGRHQAIPE-UHFFFAOYSA-N |
Synonym | samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate |
PubChem CID | 203081 |
SMILES | O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10185420
|
Thermo Scientific Acros
316150250 |
25 g | Plastic bottle |
81.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10279843
|
Thermo Scientific Acros
316151000 |
100 g | Plastic bottle |
213.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
13759-83-6 | |
444.46 | |
HDCOFJGRHQAIPE-UHFFFAOYSA-N | |
203081 |
H12N3O15Sm | |
MFCD00149857 | |
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate | |
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
13759-83-6 | |
Crystalline Powder and/or Crystals | |
H12N3O15Sm | |
MFCD00149857 | |
Solubility in water: soluble | |
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
444.46 | |
444.5 | |
Trace Metal Basis |
Yellow | |
99.9% | |
Sm(NO3)3·6H2O | |
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate | |
HDCOFJGRHQAIPE-UHFFFAOYSA-N | |
samarium(3+);trinitrate;hexahydrate | |
203081 | |
99.9% | |
Samarium(III) nitrate hexahydrate |
Safety and Handling
GHS H Statement:
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only