Learn More
Samarium(III) hydroxide hydrate, 99% (REO), Thermo Scientific Chemicals
69.60 € - 185.00 €
Chemical Identifiers
CAS | 20403-06-9 |
---|---|
Molecular Formula | H3O3Sm |
Molecular Weight (g/mol) | 201.38 |
MDL Number | MFCD00050108 |
InChI Key | BCYBEIXXOVNETJ-UHFFFAOYSA-K |
IUPAC Name | samarium(3+) trihydroxide |
SMILES | [OH-].[OH-].[OH-].[Sm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338318
|
Thermo Scientific Alfa Aesar
039512.06 |
5 g |
69.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328318
|
Thermo Scientific Alfa Aesar
039512.14 |
25 g |
185.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Samarium hydroxide (Sm(OH)3) is an important samarium compound with many potential applications in electronics, optics and catalysis. It is used as a chemical intermediate. It is used in the separation and preconcentration procedures for the determination of lead using spectrometric techniques. It has specialized uses in glass, phosphors, lasers, and thermoelectric devices and it is mainly applied in Neutron absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20403-06-9 | |
201.38 | |
BCYBEIXXOVNETJ-UHFFFAOYSA-K | |
[OH-].[OH-].[OH-].[Sm+3] |
H3O3Sm | |
MFCD00050108 | |
samarium(3+) trihydroxide |
Specifications
20403-06-9 | |
H3O3Sm | |
UN3262 | |
BCYBEIXXOVNETJ-UHFFFAOYSA-K | |
samarium(3+) trihydroxide | |
201.37 (Anhydrous) | |
(REO) |
Lump/Powder | |
MFCD00050108 | |
Soluble in water and moderately soluble in strong mineral acids. | |
[OH-].[OH-].[OH-].[Sm+3] | |
201.38 | |
99% | |
Samarium(III) hydroxide hydrate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 243-787-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only