Learn More
Samarium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals
105.53 € - 297.48 €
Chemical Identifiers
CAS | 17829-86-6 |
---|---|
Molecular Formula | C6H9O6Sm |
Molecular Weight (g/mol) | 327.49 |
MDL Number | MFCD00150124 |
InChI Key | JPDBEEUPLFWHAJ-UHFFFAOYSA-K |
IUPAC Name | samarium(3+) triacetate |
SMILES | [Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392558
|
Thermo Scientific Alfa Aesar
014604.14 |
25 g |
105.53 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382558
|
Thermo Scientific Alfa Aesar
014604.22 |
100 g |
297.48 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Samarium Acetate has specialized uses in glass, phosphors, lasers, and thermoelectric devices. Another important application of Samarium and its compounds is as catalyst and chemical reagent. One of the most important applications of Samarium is in Samarium-Cobalt magnets. . These magnets are found in small motors, headphones, and high-end magnetic pickups for guitars and related musical instruments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17829-86-6 | |
327.49 | |
JPDBEEUPLFWHAJ-UHFFFAOYSA-K | |
[Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H9O6Sm | |
MFCD00150124 | |
samarium(3+) triacetate |
Specifications
300°C | |
Crystalline Aggregates | |
MFCD00150124 | |
JPDBEEUPLFWHAJ-UHFFFAOYSA-K | |
samarium(3+) triacetate | |
327.48 (Anhydrous) | |
Odorless | |
1.94 g/mL | |
Samarium(III) acetate hydrate |
17829-86-6 | |
C6H9O6Sm | |
Soluble in water | |
[Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
327.49 | |
99.9% | |
Hygroscopic | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 233-950-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only