missing translation for 'onlineSavingsMsg'
Learn More

Salicylamidoxime, 97%, Thermo Scientific™

Product Code. 15411058
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6005-58-9
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD08061199
InChI Key DWGZGKKDBMJXGG-ALCCZGGFSA-N
Synonym salicylamidoxime, 2-hydroxy-benzamide oxime, 2,n-dihydroxy-benzamidine, 2-hydroxybenzamidoxime, z-n',2-dihydroxybenzimidamide, n',2-dihydroxybenzene-1-carboximidamide, 6z-6-amino hydroxyamino methylidene cyclohexa-2,4-dien-1-one, 6z-6-amino-hydroxyamino methylidene-1-cyclohexa-2,4-dienone, 6z-6-azanyl-oxidanylamino methylidene cyclohexa-2,4-dien-1-one
PubChem CID 20398791
IUPAC Name (6Z)-6-[amino-(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C(N)NO)C(=O)C=C1

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Salicylamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'