missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde hydrazone, 98%, Thermo Scientific Chemicals

Product Code. 11479927
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3291-00-7
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007626
InChI Key RHPIFAMEPPCHGX-AATRIKPKSA-N
Synonym salicylaldehyde hydrazone, benzaldehyde, 2-hydroxy-, hydrazone, 2-methanehydrazonoylphenol, salicylic aldehyde hydrazone, 1-salicylidenehydrazine, 6-hydrazinylmethylidene cyclohexa-2,4-dien-1-one, acmc-1ap22, myunwhtzyxucik-uhfffaoysa-n, benzaldehyde,2-hydroxy-, hydrazone
PubChem CID 6746399
SMILES NN\C=C1/C=CC=CC1=O

Specifications

Melting Point 95°C to 98°C
Quantity 25 g
IUPAC Name 6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material Salicylaldehyde hydrazone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'