Learn More
Salicylaldehyde, 99%, Thermo Scientific Chemicals
33.57 € - 286.43 €
Chemical Identifiers
CAS | 90-02-8 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.12 |
MDL Number | MFCD00003317 |
InChI Key | SMQUZDBALVYZAC-UHFFFAOYSA-N |
Synonym | salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy |
PubChem CID | 6998 |
ChEBI | CHEBI:16008 |
IUPAC Name | 2-hydroxybenzaldehyde |
SMILES | OC1=CC=CC=C1C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454924
|
Thermo Scientific Alfa Aesar
A13833.22 |
100 g |
33.57 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464924
|
Thermo Scientific Alfa Aesar
A13833.36 |
500 g |
79.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474924
|
Thermo Scientific Alfa Aesar
A13833.0E |
2500 g |
286.43 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPrecursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.
Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.
Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
Chemical Identifiers
90-02-8 | |
122.12 | |
SMQUZDBALVYZAC-UHFFFAOYSA-N | |
6998 | |
2-hydroxybenzaldehyde |
C7H6O2 | |
MFCD00003317 | |
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy | |
CHEBI:16008 | |
OC1=CC=CC=C1C=O |
Specifications
90-02-8 | |
1.168 | |
76°C (168°F) | |
C7H6O2 | |
MFCD00003317 | |
Air and light sensitive | |
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy | |
SMQUZDBALVYZAC-UHFFFAOYSA-N | |
2-hydroxybenzaldehyde | |
6998 | |
122.12 | |
Liquid |
°C to 7°C | |
197°C | |
99% | |
1.573 | |
471388 | |
14,8326 | |
Miscible with alcohol,ether,acetone,benzene and most organic solvents. Slightly miscible with water. | |
OC1=CC=CC=C1C=O | |
122.12 | |
CHEBI:16008 | |
99% | |
Salicylaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H302-H311-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 201-961-0
missing translation for 'rtecsNumber' : VN5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Salicylaldehyde
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Salicylaldehyde, phenol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.