missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ S3™ B-44-4X Tubing

Food and beverage dispensing Tygon™ S3™ B-44-4X Tubing.

137.00 €

Specifications

Product Type Tubing
Material Tygon E-Food (B-44-4X)
Color Clear
View More Specs

Products 1
Product Code Brand Flexibility Diameter (English) Inner Diameter (English) Outer Price Quantity & Availability  
Product Code Brand Flexibility Diameter (English) Inner Diameter (English) Outer Price Quantity & Availability  
15705103
View Documents Promotion Details
Saint Gobain Life Sciences
AAA00017
25 mm bend radius 1/4 in. 0.375 in.
137.00 €
15m
View alternative products
Save Up to 
»
Estimated Shipment: 15-10-2024
to see stock.
Add to basket
 
Description

Description

Tygon S3™ B-44-4X tubing from Saint-Gobain is lightweight and extremely flexible, readily accommodating abrupt corners and obstacle with ease. Compared to rigid piping, it has a very small footprint, is simple to install and only requires minimal couplings and fittings. Its glass-like clarity makes it easy to monitor and adjust the flow of foods or beverages during processing, and its smooth, non-porous bore inhibits particle entrapment, minimizing the potential for bacterial growth. Our food and beverage dispensing tubing is extremely resistant to harsh alkaline cleaners and sanitizers, and is ideal for aseptic filling, condiment dispensing, soft-serve dispensing, dairy processing and vitamin and flavor concentrate systems

  • Smooth, non-porous bore will not trap particulates or promote bacterial growth
  • Resistant to harsh alkaline cleaners* and sanitizers
  • Excellent alternative to rigid piping systems
  • Contains no BPA or phthalates

Typical Applications*

  • Aseptic filling
  • Condiment dispensing
  • Dairy processing
  • Vitamin and flavor concentrate systems
  • Soft-serve dispensing

Compliance

FDA, NSF, 3-A, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU

Specifications

Specifications

Tubing
Tygon E-Food (B-44-4X)
Clear
Videos
SDS
Documents

Documents

Product Certifications