missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers

Product Code. 12336988
Click to view available options
Capacity (Metric):
1 mL
3 mL
5 mL
10 mL
30 mL
50 mL
100 mL
250 mL
400 mL
600 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.

  • Nonwettable, unbreakable and heat resistant
  • Easy to clean
  • Broad spout for easy pouring
  • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
  • Specially molded bottom allows rapid heat transfer
  • Unlike most PTFE beakers, these will boil water when covered

Applications:

Ideal for digestion

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 33.81 oz.
Capacity (Metric) 1000 mL
Material PTFE
For Use With (Application) Ideal for Digestion
Height (English) 5.75 in.
Height (Metric) 14.62 cm
Includes Spout
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C
Type Beaker
Style Griffin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.