Learn More
Thermo Scientific Chemicals Safranin O, 85%, pure, certified, Thermo Scientific Chemicals
42.64 € - 109.55 €
Chemical Identifiers
CAS | 477-73-6 |
---|---|
Molecular Formula | C20H19ClN4 |
Molecular Weight (g/mol) | 350.85 |
MDL Number | MFCD00011759 |
InChI Key | QRYAEWIQIBAZOJ-UHFFFAOYSA-N |
Synonym | Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride |
PubChem CID | 2723800 |
IUPAC Name | 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride |
SMILES | [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055960
|
Thermo Scientific Chemicals
419210250 |
25 g | Glass bottle |
42.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444315
|
Thermo Scientific Chemicals
419211000 |
100 g | Glass bottle |
109.55 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
477-73-6 | |
350.85 | |
QRYAEWIQIBAZOJ-UHFFFAOYSA-N | |
2723800 | |
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 |
C20H19ClN4 | |
MFCD00011759 | |
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride | |
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride |
Specifications
477-73-6 | |
80% min. | |
C20H19ClN4 | |
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride | |
Authentic | |
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride | |
2723800 | |
350.84 | |
Brown to Green | |
Safranine O, Pure |
1.1 to 1.32 | |
85% | |
MFCD00011759 | |
QRYAEWIQIBAZOJ-UHFFFAOYSA-N | |
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 | |
350.85 | |
530 to 534nm | |
Pure | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Safranine O
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.