missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
Description
- Chemical name and formula
- Full color NFPA™ Diamond
- Health hazards
- CAS # (Chemical Abstract Services #)
- Suggested protective clothing and equipment
- Wide mouth for easy filling
- Color caps enhance identification
- LDPE bottles with polypropylene valves
- 500mL
- “Label Your Own” bottles also available
- Use only with solvents compatible with LDPE
Specifications
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Green |
For Use With (Application) | Methanol |
Material | LDPE |
Quantity | 4/PK |
Closure Color | Green |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Autoclavable | Not Autoclavable |
Mouth | Wide |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction