missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ PFE Latex Ambidextrous Gloves

Kimtech™ PFE Latex Ambidextrous Gloves provide comfortable, skin-friendly and ISO-compliant contamination protection. The non-sterile PFE latex gloves also feature a textured finish and beaded cuff to add strength and simplify donning.

Specifications

Certifications/Compliance EN455-1 and EN455-2, Manufactured in accordance with Quality System ISO 9001
Classification PPE Category III
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
View More Specs

Description

Description

  • Assured Compliance:
    • PPE Cat III according to Regulation (EU) 2016/425
    • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
    • EN 374-4:2003 Resistance to degradation by chemicals
    • EN ISO 374-5:2016 Micro Organism and Virus Protection
  • Superb quality PFE-latex material provides high levels of contamination protection against micro-organisms, chemical splash, viruses and more.
  • Suitable for use in a range of laboratories environments and provided with supporting technical information to demonstrate regulatory compliance.
  • Gloves are non-sterile, ambidextrous, disposable and a natural color.
  • Textured palm and fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Made from natural rubber latex for a low dermatitis risk and containing no powder, reducing the risks of skin irritation for the wearer.
Specifications

Specifications

EN455-1 and EN455-2, Manufactured in accordance with Quality System ISO 9001
Natural
Fully Textured
Yes
Latex
Powder Free
Beaded Cuff
For laboratory and safety environment
100 Pack
PPE Category III
Single-use
Ambidextrous
No
0.13 mm
Examination Gloves
Textured
Beaded/Rolled
Videos
SDS
Documents

Documents

Product Certifications